accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2388-10-5

IUPAC Name: lithium(1+) propan-2-olate
Synonyms: lithium(1+) propan-2-olate
Molecular Weight (g/mol): 66.03
Molecular Formula: C3H7LiO
InChi Key: HAUKUGBTJXWQMF-UHFFFAOYSA-N
SMILES: [Li+].CC(C)[O-]
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results