accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COOHN(S)OOCH3NH2HCl

CAS RN 2389-48-2

IUPAC Name: methyl (2S)-2-amino-6-{[(tert-butoxy)carbonyl]amino}hexanoate hydrochloride
Synonyms: methyl (2S)-2-amino-6-[(tert-butoxycarbonyl)amino]hexanoate hydrochloride
Molecular Weight (g/mol): 296.79
Molecular Formula: C12H25ClN2O4
InChi Key: NANRHOPPXCBHGI-MVRPGTNWNA-N
SMILES: Cl.COC(=O)[C@@H](N)CCCCNC(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)