accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONH(S)NHOOOHO

CAS RN 2389-60-8

IUPAC Name: (2S)-2-{[(benzyloxy)carbonyl]amino}-6-{[(tert-butoxy)carbonyl]amino}hexanoic acid
Synonyms: (2S)-2-{[(benzyloxy)carbonyl]amino}-6-[(tert-butoxycarbonyl)amino]hexanoic acid
Molecular Weight (g/mol): 380.44
Molecular Formula: C19H28N2O6
InChi Key: DYSBKEOCHROEGX-GGYSOQFKNA-N
SMILES: CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results