accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2390-60-5

ClCH3HNCNH3CCH3NH3CCH3

CAS RN 2390-60-5

IUPAC Name: bis[4-(diethylamino)phenyl][4-(ethylamino)naphthalen-1-yl]methylium chloride
Synonyms: victoria blue BO
Molecular Weight (g/mol): 514.15
Molecular Formula: C33H40ClN3
InChi Key: ZQTYUKTXBQYBSD-UHFFFAOYSA-M
SMILES: [Cl-].CCNC1=C2C=CC=CC2=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC
Color 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)