accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CSSBOOH3CH3CCH3H3C

CAS RN 239075-02-6

IUPAC Name: 4,4,5,5-tetramethyl-2-[5'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[2,2'-bithiophen]-5-yl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[5'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[2,2'-bithiophen]-5-yl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 418.18
Molecular Formula: C20H28B2O4S2
InChi Key: XWWXVHGWYCXJCJ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(S1)C1=CC=C(S1)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results