accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23912-79-0

IUPAC Name: 5,7,12,14-tetrahydropentacene-5,7,12,14-tetrone
Synonyms: pentacene-5,7,12,14-tetrone
Molecular Weight (g/mol): 338.32
Molecular Formula: C22H10O4
InChi Key: YZOGOBWHTVNKGA-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2C(=O)C2=CC3=C(C=C12)C(=O)C1=CC=CC=C1C3=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results