accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNCS

CAS RN 2392-68-9

IUPAC Name: 1-chloro-3-isothiocyanatobenzene
Synonyms: 1-chloro-3-isothiocyanatobenzene
Molecular Weight (g/mol): 169.63
Molecular Formula: C7H4ClNS
InChi Key: WGXCKFMVBAOIFH-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(=C1)N=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results