accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNHOClClCH

CAS RN 23950-58-5

IUPAC Name: 3,5-dichloro-N-(2-methylbut-3-yn-2-yl)benzamide
Synonyms: propyzamide pronamide
Molecular Weight (g/mol): 256.13
Molecular Formula: C12H11Cl2NO
InChi Key: PHNUZKMIPFFYSO-UHFFFAOYSA-N
SMILES: CC(C)(NC(=O)C1=CC(Cl)=CC(Cl)=C1)C#C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results