accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONN(E/Z)

CAS RN 2396-60-3

IUPAC Name: (4-methoxyphenyl)(phenyl)diazene
Synonyms: (4-methoxyphenyl)phenyldiazene
Molecular Weight (g/mol): 212.25
Molecular Formula: C13H12N2O
InChi Key: LGCRPKOHRIXSEG-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N=NC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)