accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3(E)

CAS RN 2396-78-3

IUPAC Name: methyl (3E)-hex-3-enoate
Synonyms: methyl (3E)-hex-3-enoate
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: XEAIHUDTEINXFG-SNAWJCMRSA-N
SMILES: CC\C=C\CC(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results