accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2396-83-0

IUPAC Name: ethyl (3E)-hex-3-enoate
Synonyms: ethyl trans-3-hexenoate
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: VTSFIPHRNAESED-AATRIKPKSA-N
SMILES: CCOC(=O)C\C=C\CC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results