accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3C(E)(E)

CAS RN 2396-84-1

IUPAC Name: ethyl (2E,4E)-hexa-2,4-dienoate
Synonyms: ethyl (2E,4E)-hexa-2,4-dienoate
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H12O2
InChi Key: OZZYKXXGCOLLLO-TWTPFVCWSA-N
SMILES: CCOC(=O)\C=C\C=C\C
Percent Purity 
  • (3)
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (5)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (5)