accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23967-57-9

OOSCl

CAS RN 23967-57-9

IUPAC Name: 4-chloro-1-benzothiophene-2-carboxylate
Synonyms: 4-chloro-1-benzothiophene-2-carboxylate
Molecular Weight (g/mol): 211.64
Molecular Formula: C9H4ClO2S
InChi Key: IPAXPERGAMNMIJ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC2=C(Cl)C=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results