accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOCH3N(E)

CAS RN 2397-00-4

IUPAC Name: 5-methyl-2-{4-[(1E)-2-[4-(5-methyl-1,3-benzoxazol-2-yl)phenyl]ethenyl]phenyl}-1,3-benzoxazole
Synonyms: 5-methyl-2-{4-[(1E)-2-[4-(5-methyl-1,3-benzoxazol-2-yl)phenyl]ethenyl]phenyl}-1,3-benzoxazole
Molecular Weight (g/mol): 442.52
Molecular Formula: C30H22N2O2
InChi Key: OKEZAUMKBWTTCR-AATRIKPKSA-N
SMILES: CC1=CC=C2OC(=NC2=C1)C1=CC=C(\C=C\C2=CC=C(C=C2)C2=NC3=CC(C)=CC=C3O2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results