accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2398-37-0

IUPAC Name: 1-bromo-3-methoxybenzene
Synonyms: 1-bromo-3-methoxybenzene
Molecular Weight (g/mol): 187.04
Molecular Formula: C7H7BrO
InChi Key: PLDWAJLZAAHOGG-UHFFFAOYSA-N
SMILES: COC1=CC=CC(Br)=C1
Boiling Point 
  • (3)
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (7)
Percent Purity 
  • (4)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (7)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)