accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3NN

CAS RN 23996-25-0

IUPAC Name: 3-(2-ethyl-4-methyl-1H-imidazol-1-yl)propanenitrile
Synonyms: 3-(2-ethyl-4-methylimidazol-1-yl)propanenitrile
Molecular Weight (g/mol): 163.22
Molecular Formula: C9H13N3
InChi Key: UIDDPPKZYZTEGS-UHFFFAOYSA-N
SMILES: CCC1=NC(C)=CN1CCC#N
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)