accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH2H2N

CAS RN 2400-78-4

IUPAC Name: 2-methylpropane-1,3-diamine
Synonyms: 2-methylpropane-1,3-diamine
Molecular Weight (g/mol): 88.15
Molecular Formula: C4H12N2
InChi Key: BDXGMDGYOIWKIF-UHFFFAOYSA-N
SMILES: CC(CN)CN
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results