accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 240131-46-8

IUPAC Name: magnesium(2+) bis(5,5,5-trifluoro-4-oxopent-2-en-2-olate) dihydrate
Synonyms: magnesium(2+) bis(5,5,5-trifluoro-4-oxopent-2-en-2-olate) dihydrate
Molecular Weight (g/mol): 366.5
Molecular Formula: C10H12F6MgO6
InChi Key: OWJDZQKGGDMWCY-UHFFFAOYSA-L
SMILES: O.O.[Mg++].CC([O-])=CC(=O)C(F)(F)F.CC([O-])=CC(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results