accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24016-03-3

IUPAC Name: 3-(benzyloxy)pyridin-2-amine
Synonyms: 3-(benzyloxy)pyridin-2-amine
Molecular Weight (g/mol): 200.24
Molecular Formula: C12H12N2O
InChi Key: NMCBWICNRJLKKM-UHFFFAOYSA-N
SMILES: NC1=NC=CC=C1OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results