accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2402-99-5

IUPAC Name: 3,5-dibromopyridin-1-ium-1-olate
Synonyms: 3,5-dibromopyridin-1-ium-1-olate
Molecular Weight (g/mol): 252.89
Molecular Formula: C5H3Br2NO
InChi Key: QBUQQUOGXPOBAQ-UHFFFAOYSA-N
SMILES: [O-][N+]1=CC(Br)=CC(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results