accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COHH3CH3CHN

CAS RN 2403-88-5

IUPAC Name: 2,2,6,6-tetramethylpiperidin-4-ol
Synonyms: 2,2,6,6-tetramethylpiperidin-4-ol
Molecular Weight (g/mol): 157.26
Molecular Formula: C9H19NO
InChi Key: VDVUCLWJZJHFAV-UHFFFAOYSA-N
SMILES: CC1(C)CC(O)CC(C)(C)N1
Molecular Weight (g/mol) 
  • (2)
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (6)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (2)
  • (5)
  • (3)
  • (2)
  • (5)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (6)
  • (2)
  • (2)
  • (3)