accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2403-89-6

IUPAC Name: 1,2,2,6,6-pentamethylpiperidin-4-ol
Synonyms: 1,2,2,6,6-pentamethylpiperidin-4-ol
Molecular Weight (g/mol): 171.28
Molecular Formula: C10H21NO
InChi Key: NWHNXXMYEICZAT-UHFFFAOYSA-N
SMILES: CN1C(C)(C)CC(O)CC1(C)C
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)