accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN(R)ONO(R)OOO(R)O

CAS RN 240408-78-0

IUPAC Name: tris({[(2R)-oxiran-2-yl]methyl})-1,3,5-triazinane-2,4,6-trione
Synonyms: tris[(2R)-oxiran-2-ylmethyl]-1,3,5-triazinane-2,4,6-trione
Molecular Weight (g/mol): 297.27
Molecular Formula: C12H15N3O6
InChi Key: OUPZKGBUJRBPGC-IWSPIJDZSA-N
SMILES: O=C1N(C[C@@H]2CO2)C(=O)N(C[C@@H]2CO2)C(=O)N1C[C@@H]1CO1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results