CAS RN 240417-00-9
IUPAC Name:
2'-(diphenylphosphanyl)-N,N-dimethyl-[1,1'-biphenyl]-2-amine
Synonyms:
2'-(diphenylphosphanyl)-N,N-dimethyl-[1,1'-biphenyl]-2-amine
Molecular Weight (g/mol):
381.46
Molecular Formula:
C26H24NP
InChi Key:
JGFXUYLYPITYGR-UHFFFAOYSA-N
SMILES:
CN(C)C1=CC=CC=C1C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Sigma Aldrich (S,S)-Dipamp
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.