accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CSOONCH3OOCH3CH3

CAS RN 240423-74-9

IUPAC Name: 2-(N-benzyl2,4,6-trimethylbenzenesulfonamido)-1-phenylpropyl acetate
Synonyms: 2-(N-benzyl2,4,6-trimethylbenzenesulfonamido)-1-phenylpropyl acetate
Molecular Weight (g/mol): 465.61
Molecular Formula: C27H31NO4S
InChi Key: MCELVTKIIIBSDU-UHFFFAOYNA-N
SMILES: CC(C(OC(C)=O)C1=CC=CC=C1)N(CC1=CC=CC=C1)S(=O)(=O)C1=C(C)C=C(C)C=C1C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results