accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOSCl

CAS RN 24065-33-6

IUPAC Name: 5-chlorothiophene-2-carboxylic acid
Synonyms: 5-chlorothiophene-2-carboxylic acid
Molecular Weight (g/mol): 162.59
Molecular Formula: C5H3ClO2S
InChi Key: QZLSBOVWPHXCLT-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(Cl)S1
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)