accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24067-17-2

IUPAC Name: (4-nitrophenyl)boronic acid
Synonyms: 4-nitrophenylboronic acid
Molecular Weight (g/mol): 166.93
Molecular Formula: C6H6BNO4
InChi Key: NSFJAFZHYOAMHL-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (6)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)