CAS RN 2407-11-6
CAS RN 2407-11-6
IUPAC Name:
2-chloro-6-nitro-1,3-benzothiazole
Synonyms:
2-chloro-6-nitro-1,3-benzothiazole
Molecular Weight (g/mol):
214.62
Molecular Formula:
C7H3ClN2O2S
InChi Key:
KUCSJGBXJBQHNI-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C2N=C(Cl)SC2=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals 2-Chloro-6-nitrobenzothiazole, 95%
CAS: 2407-11-6 MDL Number: MFCD00022852