accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrO

CAS RN 24078-12-4

IUPAC Name: 4-bromo-2-methylbenzaldehyde
Synonyms: 4-bromo-2-methylbenzaldehyde
Molecular Weight (g/mol): 199.05
Molecular Formula: C8H7BrO
InChi Key: RCBPVESMGNZMSG-UHFFFAOYSA-N
SMILES: CC1=C(C=O)C=CC(Br)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results