accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24078-21-5

IUPAC Name: methyl 3-methyl-4-nitrobenzoate
Synonyms: methyl 3-methyl-4-nitrobenzoate
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: IEFONJKJLZFGKQ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C(C)=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)