accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3O

CAS RN 2408-37-9

IUPAC Name: 2,2,6-trimethylcyclohexan-1-one
Synonyms: 2,2,6-trimethylcyclohexan-1-one
Molecular Weight (g/mol): 140.23
Molecular Formula: C9H16O
InChi Key: ZPVOLGVTNLDBFI-UHFFFAOYNA-N
SMILES: CC1CCCC(C)(C)C1=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)