accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2408-70-0

IUPAC Name: 2,4,6-tribromopyridine
Synonyms: 2,4,6-tribromopyridine
Molecular Weight (g/mol): 315.79
Molecular Formula: C5H2Br3N
InChi Key: WALXYTCBNHJWER-UHFFFAOYSA-N
SMILES: BrC1=CC(Br)=NC(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results