CAS RN 240800-46-8
CAS RN 240800-46-8
IUPAC Name:
2,3-bis(3-fluorophenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Synonyms:
2,3-bis(3-fluorophenyl)-5-phenyl-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular Weight (g/mol):
370.79
Molecular Formula:
C19H13ClF2N4
InChi Key:
ADEOAPBIWHHTEL-UHFFFAOYSA-M
SMILES:
[Cl-].FC1=CC=CC(=C1)N1N=C(N=[N+]1C1=CC(F)=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
2,3-Bis(3-fluorophenyl)-5-phenyltetrazolium Chloride 98.0+%, TCI America™
CAS: 240800-46-8 Molecular Formula: C19H13ClF2N4 Molecular Weight (g/mol): 370.788 MDL Number: MFCD00059998 InChI Key: ADEOAPBIWHHTEL-UHFFFAOYSA-M PubChem CID: 2736029 IUPAC Name: 2,3-bis(3-fluorophenyl)-5-phenyltetrazol-2-ium;chloride SMILES: C1=CC=C(C=C1)C2=NN([N+](=N2)C3=CC(=CC=C3)F)C4=CC(=CC=C4)F.[Cl-]