accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 24083-13-4

IUPAC Name: 4-(octyloxy)benzaldehyde
Synonyms: 4-(octyloxy)benzaldehyde
Molecular Weight (g/mol): 234.34
Molecular Formula: C15H22O2
InChi Key: KVOWZHASDIKNFK-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=CC=C(C=O)C=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)