accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24131-31-5

IUPAC Name: [3,5-bis(benzyloxy)phenyl]methanol
Synonyms: [3,5-bis(benzyloxy)phenyl]methanol
Molecular Weight (g/mol): 320.39
Molecular Formula: C21H20O3
InChi Key: MHHXKZKHZMSINU-UHFFFAOYSA-N
SMILES: OCC1=CC(OCC2=CC=CC=C2)=CC(OCC2=CC=CC=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results