accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24131-32-6

IUPAC Name: 1,3-bis(benzyloxy)-5-(bromomethyl)benzene
Synonyms: 1,3-bis(benzyloxy)-5-(bromomethyl)benzene
Molecular Weight (g/mol): 383.29
Molecular Formula: C21H19BrO2
InChi Key: WGMYJGAUAQXYFQ-UHFFFAOYSA-N
SMILES: BrCC1=CC(OCC2=CC=CC=C2)=CC(OCC2=CC=CC=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results