CAS RN 2415-09-0
CAS RN 2415-09-0
IUPAC Name:
3-(1-hydroxyethyl)phenol
Synonyms:
3-(1-hydroxyethyl)phenol
Molecular Weight (g/mol):
138.17
Molecular Formula:
C8H10O2
InChi Key:
COJRWHSKVYUZHQ-UHFFFAOYNA-N
SMILES:
CC(O)C1=CC(O)=CC=C1
1
–
1
of
1
results
1
3-Hydroxy-alpha-methylbenzyl Alcohol 98.0+%, TCI America™
CAS: 2415-09-0 Molecular Formula: C8H10O2 Molecular Weight (g/mol): 138.166 MDL Number: MFCD00238621 InChI Key: COJRWHSKVYUZHQ-UHFFFAOYSA-N Synonym: 1-(3-Hydroxyphenyl)ethanol, 3-(1-Hydroxyethyl)phenol PubChem CID: 13542886 IUPAC Name: 3-(1-hydroxyethyl)phenol SMILES: CC(C1=CC(=CC=C1)O)O