accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNCH3

CAS RN 2415-85-2

IUPAC Name: N-(4-methylphenyl)-3-oxobutanamide
Synonyms: N-(4-methylphenyl)-3-oxobutanamide
Molecular Weight (g/mol): 191.23
Molecular Formula: C11H13NO2
InChi Key: MJGLMEMIYDUEHA-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)NC1=CC=C(C)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results