accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24160-53-0

IUPAC Name: (2E)-3-(2,4,5-trimethoxyphenyl)prop-2-enoic acid
Synonyms: (2E)-3-(2,4,5-trimethoxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol): 238.24
Molecular Formula: C12H14O5
InChi Key: XCEGAEUDHJEYRY-SNAWJCMRSA-N
SMILES: COC1=CC(OC)=C(\C=C\C(O)=O)C=C1OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results