accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24168-70-5

IUPAC Name: 2-(butan-2-yl)-3-methoxypyrazine
Synonyms: 3-tert-butyl-2-methoxypyrazine
Molecular Weight (g/mol): 166.22
Molecular Formula: C9H14N2O
InChi Key: QMQDJVIJVPEQHE-UHFFFAOYNA-N
SMILES: CCC(C)C1=NC=CN=C1OC
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results