accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CHOCH3OHCH3

CAS RN 2417-04-1

IUPAC Name: 3,3',5,5'-tetramethyl-[1,1'-biphenyl]-4,4'-diol
Synonyms: 3,3',5,5'-tetramethyl-[1,1'-biphenyl]-4,4'-diol
Molecular Weight (g/mol): 242.32
Molecular Formula: C16H18O2
InChi Key: YGYPMFPGZQPETF-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1O)C1=CC(C)=C(O)C(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results