accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2417-72-3

IUPAC Name: methyl 4-(bromomethyl)benzoate
Synonyms: methyl 4-(bromomethyl)benzoate
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: NLWBJPPMPLPZIE-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(CBr)C=C1
Percent Purity 
  • (2)
  • (8)
Quantity 
  • (1)
  • (2)
  • (4)
  • (3)
Boiling Point 
  • (5)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (2)
  • (8)
  • (1)
  • (2)
  • (4)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)