accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24171-03-7

IUPAC Name: 2,5-diaminobenzene-1,4-diol
Synonyms: 2,5-diaminobenzene-1,4-diol
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: RLXBOUUYEFOFSW-UHFFFAOYSA-N
SMILES: NC1=CC(O)=C(N)C=C1O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)