accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2419-56-9

IUPAC Name: 2-amino-5-(tert-butoxy)-5-oxopentanoic acid
Synonyms: 2-amino-5-(tert-butoxy)-5-oxopentanoic acid
Molecular Weight (g/mol): 203.24
Molecular Formula: C9H17NO4
InChi Key: OIOAKXPMBIZAHL-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)CCC(N)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)