accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2419-73-0

OH(S)Cl(R)OHCl

CAS RN 2419-73-0

IUPAC Name: (2R,3S)-1,4-dichlorobutane-2,3-diol
Synonyms: (2R,3S)-1,4-dichlorobutane-2,3-diol
Molecular Weight (g/mol): 159.01
Molecular Formula: C4H8Cl2O2
InChi Key: SAUBRJOIKMVSRU-ZXZARUISSA-N
SMILES: O[C@@H](CCl)[C@H](O)CCl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)