accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClCl

CAS RN 2419-74-1

IUPAC Name: 1,4-dichlorobutan-2-ol
Synonyms: 1,4-dichloro-2-hydroxybutane
Molecular Weight (g/mol): 143.01
Molecular Formula: C4H8Cl2O
InChi Key: CKNNDWZSFAPUJS-UHFFFAOYNA-N
SMILES: OC(CCl)CCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results