accessibility menu, dialog, popup

UserName

Viewing profile as user:
SSSS

CAS RN 24194-61-4

IUPAC Name: 1,4,8,11-tetrathiacyclotetradecane
Synonyms: 1,4,8,11-tetrathiacyclotetradecane
Molecular Weight (g/mol): 268.51
Molecular Formula: C10H20S4
InChi Key: WMVHWZHZRYGGOO-UHFFFAOYSA-N
SMILES: C1CSCCSCCCSCCSC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results