CAS RN 2420-26-0
IUPAC Name:
4-chloro-2-hydroxybenzaldehyde
Synonyms:
4-chlorosalicylaldehyde
Molecular Weight (g/mol):
156.57
Molecular Formula:
C7H5ClO2
InChi Key:
QNZWAJZEJAOVPN-UHFFFAOYSA-N
SMILES:
OC1=C(C=O)C=CC(Cl)=C1
1
–
1
of
1
results
1
4-Chlorosalicylaldehyde 95.0+%, TCI America™
CAS: 2420-26-0 Molecular Formula: C7H5ClO2 Molecular Weight (g/mol): 156.57 MDL Number: MFCD06252499 InChI Key: QNZWAJZEJAOVPN-UHFFFAOYSA-N Synonym: 4-Chloro-2-hydroxybenzaldehyde PubChem CID: 520101 IUPAC Name: 4-chloro-2-hydroxybenzaldehyde SMILES: OC1=C(C=O)C=CC(Cl)=C1