accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2423-73-6

IUPAC Name: 5-nitro-3-phenyl-[1,1'-biphenyl]-2-ol
Synonyms: 5-nitro-3-phenyl-[1,1'-biphenyl]-2-ol
Molecular Weight (g/mol): 291.31
Molecular Formula: C18H13NO3
InChi Key: YCXQKJXTGDYKIO-UHFFFAOYSA-N
SMILES: OC1=C(C=C(C=C1C1=CC=CC=C1)[N+]([O-])=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results