accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24241-18-7

IUPAC Name: 3,5-dibromopyrazin-2-amine
Synonyms: 3,5-dibromopyrazin-2-amine
Molecular Weight (g/mol): 252.9
Molecular Formula: C4H3Br2N3
InChi Key: DTLBKXRFWUERQN-UHFFFAOYSA-N
SMILES: NC1=NC=C(Br)N=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results